missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Sulfate Hexadecahydrate, Extra Pure, SLR, Fisher Chemical™
£74.05 - £175.00
Chemical Identifiers
CAS | 16828-11-8 |
---|---|
Molecular Formula | Al2H22O20S3 |
Molecular Weight (g/mol) | 492.30 |
MDL Number | 149137 |
InChI Key | JANHUGNIVASISO-UHFFFAOYSA-N |
IUPAC Name | tris(sulfuric acid) dialuminium octahydrate |
SMILES | O.O.O.O.O.O.O.O.[Al].[Al].OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O |
Description
Chemical Identifiers
16828-11-8 | |
492.30 | |
JANHUGNIVASISO-UHFFFAOYSA-N | |
O.O.O.O.O.O.O.O.[Al].[Al].OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O |
Al2H22O20S3 | |
149137 | |
tris(sulfuric acid) dialuminium octahydrate |
Specifications
86°C | |
White | |
Solid | |
149137 | |
O.O.O.O.O.O.O.O.[Al].[Al].OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O | |
492.30 |
16828-11-8 | |
2.5 | |
Al2H22O20S3 | |
JANHUGNIVASISO-UHFFFAOYSA-N | |
tris(sulfuric acid) dialuminium octahydrate | |
342.1358g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Aluminium sulfate hexadecahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.