missing translation for 'onlineSavingsMsg'
Learn More
Aluminum bromide, 98%, Thermo Scientific Chemicals Product Code.: 11306138

Aluminum bromide, 98%, Thermo Scientific Chemicals

Product Code. 11306138
100 g
Click to view available options
Quantity:
10 g
100 g
This item is not returnable. View return policy

Product Code. 11306138

Brand: Thermo Scientific Alfa Aesar 011113.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
Synonym Aluminum tribromide
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 265°C
Melting Point 97.5°C
Color Orange to White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 98%
Solubility Information Reacts with water violently,soluble in alcohol,carbon disulfide,ether.Slightly soluble in methanol,diethyl ether and acetone.
Formula Weight 266.71
Odor Pungent
Sensitivity Moisture sensitive
Decomposition Information Reacts with water violently
Density 3.21 g/cm3
Chemical Name or Material Aluminum bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP25L017 Certificate TypeCertificate of Analysis Date27/03/2025 Catalog Number011113.09,011113.59,011113.22
Lot NumberP15H003 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number011113.09,011113.59,011113.22
Lot NumberU16I002 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog Number011113.09,011113.59,011113.22
Lot NumberL21K14 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011113.09,011113.59,011113.22
Lot NumberQ22G036 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number011113.09,011113.59,011113.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum bromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.