Learn More
Aluminum bromide, 98%, Thermo Scientific Chemicals
Description
Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 265°C |
Melting Point | 97.5°C |
Color | Orange to White |
Physical Form | Crystalline |
Quantity | 10 g |
Assay Percent Range | 98% |
Solubility Information | Reacts with water violently,soluble in alcohol,carbon disulfide,ether.Slightly soluble in methanol,diethyl ether and acetone. |
Formula Weight | 266.71 |
Odor | Pungent |
Sensitivity | Moisture sensitive |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P25L017 | Certificate of Analysis | 27/03/2025 | 011113.09,011113.59,011113.22 |
P15H003 | Certificate of Analysis | 13/10/2022 | 011113.09,011113.59,011113.22 |
U16I002 | Certificate of Analysis | 23/08/2022 | 011113.09,011113.59,011113.22 |
L21K14 | Certificate of Analysis | 12/11/2021 | 011113.09,011113.59,011113.22 |
Q22G036 | Certificate of Analysis | 12/11/2021 | 011113.09,011113.59,011113.22 |
Safety and Handling


- Aluminum bromide
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.