missing translation for 'onlineSavingsMsg'
Learn More
Aluminum bromide, 99.997% (metals basis), Thermo Scientific Chemicals Product Code.: 11349379

Aluminum bromide, 99.997% (metals basis), Thermo Scientific Chemicals

Product Code. 11349379
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11349379

Brand: Thermo Scientific Alfa Aesar 089065.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aluminum Bromide, is used as a catalyst for the Friedel-Crafts alkylation reaction, and also for bromination and isomerization in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
Synonym Aluminum tribromide
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 265°C
Melting Point 97.5°C
Color Orange to White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 99.997% (Metals basis)
Solubility Information Soluble in benzene,alcohol,carbon disulfide,ether and toluene. Reacts with water.
Formula Weight 266.71
Odor Pungent
Sensitivity Moisture sensitive
Density 3.21 g/cm3
Chemical Name or Material Aluminum bromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX01K036 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number089065.06,089065.14
Lot NumberR30J031 Certificate TypeCertificate of Analysis Date07/06/2023 Catalog Number089065.06,089065.14
Lot NumberT19H047 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number089065.06,089065.14
Lot NumberG22P18 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number089065.06,089065.14
Lot NumberI09Z017 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number089065.06,089065.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum bromide, 99.997% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.