Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Aluminum RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand:  Sperian™ 1785014
 View more versions of this product
Product Code. 11856950
  • £61.98 / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Body made of aluminum provides optimum resistance to hazards
  • Low breathing resistance
  • Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Canister
RD40
Aluminum alloy with PE cover activated and impregnated charcoal.
Respiratory protection against organic gases and vapors with a boiling point below 65°C, inorganic gases and vapors, solid and liquid particles
Model AXB2P3
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 0070-003-079-06-96-0120 Ext. 01-02-00, EC Attestation- EC Attestation, EC Attestation Number- 0070-003-079-06-96-0120 Ext. 01-02-00, Declaration
Gray, White, Green
ALU RD40 AXBP3
Full-Face mask equipped with an Rd40 connector.
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sperian™ Honeywell™ Aluminum RD40 Filters > Model: AXB2P3