Learn More
Heidolph™ Receiving Flask
For use with rotary evaporators
Brand:  Heidolph™ 514-83000-00
 View more versions of this product
Product Code. 10660113
  • £73.75 / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With S 35/20
  • Part number 15-300-004-11: includes drain valves
    • Included as a standard for all Hei-VAP Industrial models
Order Info
CapacityMetric
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Receiving Flask
35/20
Glass
Rotary evaporators
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph™ Receiving Flask > Capacity: 500 mL