Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Aluminum RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand: Sperian™ 1781000
This item is not returnable.
View return policy
Description
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Canister | |
RD40 | |
ALU RD40 A2B2E2K2 | |
Full-face mask equipped with an Rd40 connector | |
1 Ea. |
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 7549 A/12/23 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7549 A/12/23 PSA, Certifications- EN 14387, EN 12941, lockedHON filter | |
Aluminum alloy with PE cover activated and impregnated charcoal | |
Respiratory protection against many hazardous gases, vapors and/or particulates in many applications | |
Model A2B2E2K2RD |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Product Title
Sperian™ Honeywell™ Aluminum RD40 Filters > Model: A2B2E2K2RD
Spot an opportunity for improvement?Share a Content Correction