missing translation for 'onlineSavingsMsg'
Learn More
Aluminum chloride, 98+%, extra pure, anhydrous powder, Thermo Scientific Chemicals Product Code.: 11305015

Aluminum chloride, 98+%, extra pure, anhydrous powder, Thermo Scientific Chemicals

Product Code. 11305015
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11305015

Brand: Thermo Scientific Alfa Aesar L18489.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

Solubility
Soluble in hydrochloric acid, ethanol, chloroform and carbon tetrachloride.Slightly soluble in benzene.

Notes
Aluminum chloride is hygroscopic. It must be packed in a tightly closed container and stored in a cool, dry, and well-ventilated area. It is water-reactive and corrosive in nature. It reacts vigorously with bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminumShow MoreShow Less
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 190°C (sublimation)
pH 2.4
Physical Form Powder
Quantity 500 g
UN Number UN1726
Merck Index 14,337
Solubility Information Soluble in hydrochloric acid,ethanol,chloroform and carbon tetrachloride.Slightly soluble in benzene.
Formula Weight 133.34
Percent Purity 99%
Odor Pungent
Sensitivity Moisture sensitive
Density 2.44
Chemical Name or Material Aluminum chloride, anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX30K036 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog NumberL18489.36,L18489.22
Lot NumberA0461105 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL18489.36,L18489.22
Lot NumberS27K025 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog NumberL18489.36,L18489.22
Lot NumberR17K033 Certificate TypeCertificate of Analysis Date06/08/2024 Catalog NumberL18489.36,L18489.22
Lot NumberN18I009 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL18489.36,L18489.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum chloride, 98+%, extra pure, anhydrous powder, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.