missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Aluminum Eco AluRack for Chest Freezers
Providing a whole new way of making storage racks. Tenak™ Aluminum Eco AluRacks for Chest Freezers fit into any freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11522
This item is not returnable.
View return policy
Description
- Made of seawater resistant aluminum
- Available in a range of box heights
- Racks can be used with all freezer brands
- Sold without cryoboxes
Specifications
Eco AluRack | |
Aluminum | |
1 Pack | |
140 x 140 x 832mm | |
832 mm | |
140 mm |
Boxes (15) | |
For chest freezers | |
1 Pack | |
140 mm | |
50 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tenak™ Aluminum Eco AluRack for Chest Freezers > Holds: 15 boxes, 140 x 140 x 832mm
Spot an opportunity for improvement?Share a Content Correction