missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Aluminum Eco AluRack for Chest Freezers

Providing a whole new way of making storage racks. Tenak™ Aluminum Eco AluRacks for Chest Freezers fit into any freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11516

 View more versions of this product

Product Code. 12448120

  • £63.40 / Each
Estimated Shipment: 21-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of seawater resistant aluminum
  • Available in a range of box heights
  • Racks can be used with all freezer brands
  • Sold without cryoboxes
Specifications

Specifications

Eco AluRack
Aluminum
12
For Mtp, chest freezers
1 Pack
140 mm
140 mm
Boxes (12)
12
Yes
1 Pack
140 x 140 x 665 mm
665 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tenak™ Aluminum Eco AluRack for Chest Freezers > Holds: 12 boxes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.