Learn More
Aluminum oxide, activated, neutral, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals
Description
Aluminum oxide is used as an abrasive, dosimeter, filler of choice for plastics, ingredient in sunscreen and cosmetics. Aluminum oxide flakes are also used in paint for providing reflective decorative effects. In laboratories it is widely used as a medium for chromatographic purifications. It is used in the production of aluminum (Hall–Héroult process), alumina nanofibers, cutoff tools, and sandpaper. It serves as a catalyst for several reactions of industrial importance, including converting alcohols into alkenes, decomposing hydrazine, drying of gases, and converting hydrogen sulfide into sulfur in refineries (Claus Process). It is also used in integrated circuits, compact fluorescent lamps (CFL), sodium vapor lamps, hip replacements, CD/DVD polishing, superconducting quantum interference devices (SQUIDS), insulation devices, and as a support for many popular catalysts (for example, in Ziegler-Natta polymerizations).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2045°C |
Physical Form | Powder |
Quantity | 250 g |
Solubility Information | Insoluble in water,ethanol and diethyl ether. |
IUPAC Name | dialuminium(3+) trioxidandiide |
Formula Weight | 101.96 |
Grade | Brockmann |
Odor | Odorless |
Appearance | White powder |
Chemical Name or Material | Aluminum oxide, neutral, acidic, 58 angstroms |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M16L006 | Certificate of Analysis | 17/01/2025 | 011502.A7,011502.30,011502.A1 |
W23K040 | Certificate of Analysis | 03/10/2024 | 011502.A7,011502.30,011502.A1 |
T13J015 | Certificate of Analysis | 19/07/2023 | 011502.A7,011502,011502.30,011502.A1 |
R19J014 | Certificate of Analysis | 31/05/2023 | 011502.30 |
Q20J027 | Certificate of Analysis | 01/05/2023 | 011502.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.