Learn More
Aluminum selenide, 99% (metals basis), Thermo Scientific Chemicals
£64.80 - £984.00
Chemical Identifiers
CAS | 1302-82-5 |
---|---|
MDL Number | MFCD00014158 |
Synonym | unii-4r7pmp996q, aluminum selenide, aluminum selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide |
Description
It is used as a precursor to hydrogen selenide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1302-82-5 | |
MFCD00014158 | |
unii-4r7pmp996q, aluminum selenide, aluminum selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide |
Specifications
1302-82-5 | |
Pieces | |
UN3283 | |
unii-4r7pmp996q, aluminum selenide, aluminum selenide, dialuminum selenium 2-, dialuminium 3+ triselenide, dialuminium 3+ ion triselandiide | |
290.84 | |
Odorless | |
3.437 g/mL | |
Aluminum selenide |
Brown | |
MFCD00014158 | |
14,361 | |
Decomposes to Al203 and SeO2 in water | |
99% | |
Moisture sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
missing translation for 'einecsNumber' : 215-110-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum selenide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Precautionary Statement
- P301-IF SWALLOWED:.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P330-Rinse mouth.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P373-DO NOT fight fire when fire reaches explosives.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P410-Protect from sunlight.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only