missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets come in a variety of sizes.

Brand:  Macherey-Nagel™ 818032

74.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10786902

  • £85.65 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets come in a variety of sizes.

Alugram™ Aluminum Sheets for TLC
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75mL/g,
  • Particle size 5–17μm
  • Standard grade
Thickness
  • For analytical plates 0.25mm
  • For preparative plates 0.5 and 1mm
  • For 2mm preparative layers, a slightly coarser material is used
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications

Specifications

Aluminum Sheets Alugram SIL G
60 Å
TLC Plate
0.75 mL/g
0.2 mm
5 to 17 μm
50 Pk.
Thin Layer Chromatography (TLC)
∼500 m2/g
5 x 20 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.