missing translation for 'onlineSavingsMsg'
Learn More
Aluminum sulfide, 99+%, (metals basis), Thermo Scientific Chemicals Product Code.: 11357587

Aluminum sulfide, 99+%, (metals basis), Thermo Scientific Chemicals

Product Code. 11357587
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11357587

Brand: Thermo Scientific Alfa Aesar 012851.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1302-81-4
Molecular Formula Al2S3
Molecular Weight (g/mol) 150.14
MDL Number MFCD00014162
InChI Key COOGPNLGKIHLSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trisulfanediide
SMILES [Al+3].[Al+3].[S--].[S--].[S--]

Specifications

Melting Point 1100°C
Color Yellow
Physical Form Pieces
Quantity 5 g
Assay Percent Range >99% (Metal basis)
Formula Weight 150.16
Odor Like rotten eggs
Sensitivity Moisture sensitive
Appearance Yellowish granular
Density 2.02 g/cm3
Chemical Name or Material Aluminum sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ28C084 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012851.06,012851.14
Lot NumberX28B012 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012851.06,012851.14
Lot NumberX28B011 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012851.06,012851.14
Lot NumberP09C043 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012851.06,012851.14
Lot NumberX01B014 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012851.06,012851.14
compliance-icons compliance-icons
Product Identifier
  • Aluminum sulfide
Signal Word
  • Danger
Hazard Category
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum sulfide, 99+%, (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.