missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Hybond™ PVDF Membranes: Rolls

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

Brand:  Cytiva 10600030

406.11 GBP valid until 2025-12-31
Use promo code "24363" to get your promotional price.


 View more versions of this product

Product Code. 15249814

  • £504.00 / 1 roll
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.

p>Supplied in rolls.

  • Hydrophobic PVDF membranes
  • Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights

Amersham Hybond 0.2μm

  • Compatible with a range of solvents used for rapid destaining
  • Amersham Hybond SEQ 0.2μm
    • Specifically designed for protein sequencing applications and solid phase amino acid analysis
    • Offers excellent blotting for proteins Mr<20,000
    Amersham Hybond LFP 0.2μm
    • Specifically optimized for fluorescence detection in Western blotting applications
    • Provides low background fluorescence, resulting in high sensitivity

    TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Specifically designed for protein sequencing applications and solid phase amino acid analysis
    Roll
    4 m x 260 mm
    SEQ 0.2 μm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.