missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Hybond™ PVDF Membranes: Rolls
Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.
£374.00 - £521.00
Product Code | Brand | For Use With (Application) | Grade | Format | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Grade | Format | Price | Quantity & Availability | |||
15239814
|
Cytiva
10600021 |
Compatible with a range of solvents used for rapid destaining | 0.2 μm | Roll |
£374.00
1 roll |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||
15249814
|
Cytiva
10600030 |
Specifically designed for protein sequencing applications and solid phase amino acid analysis | SEQ 0.2 μm | Roll |
£490.00
1 roll |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
15279884
|
Cytiva
10600022 |
Specifically optimized for fluorescence detection in Western blotting applications | LFP 0.2 μm | Roll |
£521.00
1 roll |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||
Description
Ideal for Western blotting applications where stripping and reprobing are necessary. Cytiva Life Sciences™ Amersham™ Hybond™ PVDF Membranes: Rolls are hydrophobic PVDF membranes.
- Supplied in rolls.
- Hydrophobic PVDF membranes
- Small pore size eliminates blow-through and increases protein binding over a wide range of molecular weights
Amersham Hybond 0.2 μm
- Compatible with a range of solvents used for rapid destaining
Amersham Hybond SEQ 0.2 μm
- Specifically designed for protein sequencing applications and solid phase amino acid analysis
- Offers excellent blotting for proteins Mr<20,000
Amersham Hybond LFP 0.2 μm
- Specifically optimized for fluorescence detection in Western blotting applications
- Provides low background fluorescence, resulting in high sensitivity
Specifications
Roll | |
4 m x 260 mm |