missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Western Blotting Sandwiches
Precut, preassembled Protran Premium NC Nitrocellulose membrane and filter paper sandwiches save time
Brand: Cytiva 10600117
105.47 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
- Preassembled with two Whatman™ medium thickness 3MM Chr (0.34mm) filter papers to simplify set up
- Fit most precase gel formats
Specifications
8 cm | |
Pack of 10 | |
0.45 μm |
9 cm | |
Sandwich |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger||
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction