Learn More
Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls
Use for robust protein blotting. Cytiva Life Sciences™ Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Brand: Cytiva 10600018
194.14 GBP valid until 2024-12-31
Use promo code "21973" to get your promotional price.
Description
Use for robust protein blotting. Cytiva Life Sciences™ Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Rolls offer excellent sensitivity, resolution, and low background.
Roll dispensers.
- Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
- The support does not affect transfer conditions or results
Specifications
4 m | |
Each | |
Nitrocellulose, Inert Polyester |
20 cm | |
Roll | |
0.45 μm |
Safety and Handling
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
- UNKNOWN ACUTE-70 per cent of the mixture consists of component(s) of unknown acute toxicity.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.