missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Stirred Cells
Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.
Brand: Merck UFSC40001
Alert:
Pressure Limitation. Do not operate the stirred cell above 5.2 bar (75 psi).
Description
This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.Services
One year

Specifications
Autoclavable | |
Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells | |
Amicon™ Stirred Cell 400 mL | |
400 mL |
Amicon Stirred Cell 400 mL | |
Concentration, Buffer Exchange, Desalting | |
Ultrafiltration Equipment |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Merck Amicon™ Stirred Cells > 400mL
Spot an opportunity for improvement?Share a Content Correction