missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IPTG, dioxane-free

Induce the expression of genes under the control of lac operon in cloning and protein expression procedures.

Brand:  Thermo Scientific™ R0393

172.67 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


226.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10849040

  • £259.00 / 25g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon . It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.

It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal to determine the lac phenotype in blue/white colony screening.

Ready-to-use 100mM IPTG aqueous solution is non-toxic, stable, 0.22μm membrane-filtered, formulated for direct use in blue/white colony screening.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Isopropylthiogalactoside
IPTG Solution
Powder
0.1 mM
5 x 5 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2800898 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0393
Lot Number2821209 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0393
Lot Number2833820 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR0393
Lot Number2856962 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0393
Lot Number2878199 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0393
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ IPTG, dioxane-free > 5 x 5g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.