missing translation for 'onlineSavingsMsg'
Learn More
Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals Product Code.: 11342537

Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals

Product Code. 11342537
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11342537

Brand: Thermo Scientific Alfa Aesar L11632.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aminoguanidine nitrate is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aminoguanidine nitrate is used as a pharmaceutical raw material.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with reducing agents, strong oxidizing agents, organic materials and metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10308-82-4
Molecular Formula CH7N5O3
Molecular Weight (g/mol) 137.10
MDL Number MFCD00013174
InChI Key PMGFHEJUUBDCLU-UHFFFAOYSA-N
Synonym aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acidShow MoreShow Less
PubChem CID 165859
IUPAC Name 2-aminoguanidine;nitric acid
SMILES O[N+]([O-])=O.NN=C(N)N

Specifications

Melting Point 138°C to 142°C
Quantity 100 g
UN Number UN1479
Beilstein 3916056
Solubility Information Soluble in water.
Formula Weight 137.1
Percent Purity 98%
Assay cont. ≈10% water
Chemical Name or Material Aminoguanidine nitrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10223425 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL11632.22,L11632.14
Lot Number10158561 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberL11632.22,L11632.14
Lot Number10159239 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11632.22,L11632.14
Lot Number10197597 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11632.22,L11632.14
Lot Number90007682 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11632.22,L11632.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aminoguanidine nitrate, 98%, cont. ca 10% water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.