missing translation for 'onlineSavingsMsg'
Learn More
Aminoguanidine nitrate, 99%, Thermo Scientific Chemicals Product Code.: 10254510

Aminoguanidine nitrate, 99%, Thermo Scientific Chemicals

Product Code. 10254510
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10254510

Brand: Thermo Scientific Acros 103760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10308-82-4
Molecular Formula CH7N5O3
Molecular Weight (g/mol) 137.10
MDL Number MFCD00013174
InChI Key PMGFHEJUUBDCLU-UHFFFAOYSA-N
Synonym aminoguanidine nitrate, aminoguanidine, nitrate, hydrazinecarboximidamide, mononitrate, monoaminoguanidium nitrate, aminoguanidine, mononitrate, hydrazinecarboximidamide nitrate, hydrazinecarboximidamide, nitrate 1:1, 1-aminoguanidine; nitric acid, aminoguanidine; nitric acidShow MoreShow Less
PubChem CID 165859
IUPAC Name 2-aminoguanidine;nitric acid
SMILES O[N+]([O-])=O.NN=C(N)N

Specifications

Melting Point 145°C to 147°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula H2NNHC(=NH)NH2·HNO3
Quantity 5 g
Beilstein 03,117
Formula Weight 137.11
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Aminoguanidine nitrate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0347943 Certificate TypeCertificate of Analysis Date24/12/2021 Catalog Number103760050,103760250,103761000
compliance-icons compliance-icons
Product Identifier
  • Aminoguanidine nitrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aminoguanidine nitrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.