missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AminoLink™ Coupling Resin

Covalently immobilize proteins and antibodies by lysine residues or other primary amines (-NH2) via reductive amination with these aldehyde-activated agarose beads.

Brand:  Thermo Scientific™ 20382

Code : PN

Additional Details : CAS Number : 22919-26-2 Weight : 0.80000kg

 View more versions of this product

Product Code. 11836774

  • £441.00 / 50mL
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilization of antibodies and other proteins through primary amines.

AminoLink Coupling Resin is crosslinked 4% beaded agarose that has been activated with aldehyde groups. Proteins and other molecules with primary amines can be covalently attached (immobilized) to the resin to make chromatography columns for use in affinity purification. The aldehyde groups form stable secondary amine bonds with primary amines such as exist in the side chain of lysine (K) residues, which are generally abundant and readily accessible in proteins. Once a protein is immobilized, the prepared affinity resin can be used for a variety of batch and column affinity purification methods involving binding interactions with the immobilized protein. The resin and linkage are stable in most binding and elution conditions typically used in affinity chromatography, enabling prepared resin to be used for multiple rounds of affinity purification procedures.

Highlights:

  • AminoLink Coupling Resin – aldehyde-activated crosslinked 4% beaded agarose

  • Ideal for antibodies and other proteins – immobilize molecules via primary amines (-NH2)

  • Flexible coupling conditions – efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer with or without organic solvent

  • Stable, permanent immobilization – coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand

  • Better than immobilization to CNBr-activated agarose – bond is more stable and uncharged, resulting in less nonspecific binding in affinity purification procedures

  • Versatile and reusable – prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions

Includes:

Kit contains pre-packed AminoLink Columns, coupling buffer, quenching buffer, wash solution, sodium cyanoborohydride solution, and column accessories

Specifications

Specifications

AminoLink Coupling Resin
AminoLink™
Antibody, Proteins through Primary Amine
Aldehyde
Affinity, Agarose Resin
Store at 4°C.
50 mL
Resin
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.