missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Cerium(Iv) Nitrate, Honeywell Fluka™

Puriss. p.a., ACS Reagent,≥98.5% (RT)

£72.70 - £185.48

Chemical Identifiers

CAS 16774-21-3
MDL Number MFCD00151121
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15622190
View Documents
Honeywell-Fluka
22249-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15632190
View Documents
Honeywell-Fluka
22249-500G
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

16774-21-3
MFCD00151121

Specifications

16774-21-3
MFCD00151121
548.22g/mol
ACS/puriss. p.a.
Ammonium Cerium(Iv) Nitrate
Ce(NH4)2(NO3)6
3085
≥98.5% (RT)
Plastic Bottle
Videos
Safety and Handling

Safety and Handling

P210-P220-P260-P280-P305 + P351 + P338-P370 + P378

H272-H290-H302-H314-H317-H410

missing translation for 'einecsNumber' : 240-827-6

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.