missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Iron(Iii) Citrate, Honeywell™

Purum p.a., 14.5-16% Fe basis (RT), powder, green

£21.95 - £87.72

Chemical Identifiers

CAS 1185-57-5
Molecular Formula C6H11FeNO7+3
Molecular Weight (g/mol) 264.999
InChI Key FRHBOQMZUOWXQL-UHFFFAOYSA-N
Synonym ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
PubChem CID 118984355
IUPAC Name azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15630890
View Documents
Honeywell Research Chemicals
09713-50G
50 g
£21.95
50g
Please to purchase this item. Need a web account? Register with us today!
15620890
View Documents
Honeywell Research Chemicals
09713-500G
500 g
£44.32
500g
Please to purchase this item. Need a web account? Register with us today!
15610890
View Documents
Honeywell Research Chemicals
09713-1KG
1 kg
£87.72
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications

Chemical Identifiers

1185-57-5
264.999
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
C6H11FeNO7+3
FRHBOQMZUOWXQL-UHFFFAOYSA-N
118984355
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]

Specifications

1185-57-5
Powder
C6H8O7 ·xFe3+ · yNH3
FRHBOQMZUOWXQL-UHFFFAOYSA-N
azane;2-hydroxypropane-1,2,3-tricarboxylic acid;iron(3+)
118984355
14.5 to 16% Fe basis (RT)
Green
C6H11FeNO7+3
ammonium ferric citrate, ferri seltz, ammonii ferri citras, ferric ammonium citrate, iron ammonium citrate, ammonium iron 3+ citrate, ammonium iron iii citrate, iron ammonium citrate, green, ferri-ammoniumcitrat, braunes, ferriseltz
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Fe+3]
264.999
purum p.a.
Ammonium Iron(Iii) Citrate
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

H315-H319-H335

missing translation for 'einecsNumber' : 214-686-6

missing translation for 'rtecsNumber' : GE7540000

SDS
Documents

Documents

Product Certifications