missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ampule Breakers

Thermo Scientific Ampule Breakers provide a safe, convenient way to open 1 mL to 10 mL glass ampules.

£34.14 - £34.29

Specifications

Disposable Single-use
Product Type Ampule Breaker
View More Specs

Products 2
Product Code Brand Product Type Size Price Quantity & Availability  
Product Code Brand Product Type Size Price Quantity & Availability  
15940372
View Documents Promotion Details
Thermo Scientific™
28915
Ampule Breaker 1-4 mL
£34.29
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
16365212
View Documents Promotion Details
Thermo Scientific™
28916
Ampule Breaker 5-10 mL
£34.14
Pack of 100
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
Description

Description

These single-use polyethylene sleeve ampule breakers help protect your fingers from glass shards when opening 1 mL to 10 mL glass ampules. The Thermo Scientific Ampule Breakers encapsulate the ampule top upon opening, making disposal of the glass top easy and safe.

  • Easy to use single-use format
  • Compatible with 1 mL to 10 mL glass ampules
  • Protects against glass shards
  • Helps prevent cross contamination
Specifications

Specifications

Single-use
Ampule Breaker
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Ampule Breakers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.