missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Anti-Human B-Lymphocyte Value Bundle (RUO/GMP)
Monoclonal antibodies for detecting human antigens.
Brand: BD Biosciences™ 340428
Additional Details : Weight : 0.05710kg
Description
- Contents:
- CD19, CD69, CD45 antibody cocktail
- CD45 Isotype: control cocktail
- Research applications include studies of the following:
- Negative control for immunofluorescence staining of peripheral blood with three-color reagents using CD45 PerCP fluorescence triggering and gating.
Specifications
CD19, CD69, CD45 | |
Mouse | |
RUO | |
Human |
Flow Cytometry | |
50 Tests | |
Storage buffer: Phosphate buffered saline with BSA, beta-lactoglobulin, and 0.1% sodium azide. | |
Store vials at 2°C–8°C. Conjugated forms should not be frozen. Protect from exposure to light. Each reagent is stable until the expiration date shown on the bottle label when stored as directed. |