Learn More
Clark Scientific™ Polystyrene Anti-static Spill Trays
Made from anti-static, high impact polystyrene, these spill trays are ideal in most research, scientific, laboratory and other environments.
Brand:  Clark Scientific™ AT1-PK25
Product Code. 12928837
  • £258.00 / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Made from anti-static, high impact polystyrene
  • Sufficient depth to safely contain even the largest liquid spills
  • Wipe clean for repeated use
Specifications
Specifications
475 x 360 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Clark Scientific™ Polystyrene Anti-static Spill Trays > Polystyrene