Learn More
Antimony(III) bromide, 99%, Thermo Scientific Chemicals
Description
Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 280°C |
Melting Point | 96.6°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
Assay Percent Range | 99% |
Solubility Information | Soluble in alcohol,acetone,ammonia,benzene,chloroform,dilute hydrochloric acid,hydrobromic acid and carbon disulfide. |
Formula Weight | 361.48 |
Odor | Characteristic |
Refractive Index | 1.74 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y06D019 | Certificate of Analysis | 14/01/2025 | 041573.18,041573.30 |
T27I018 | Certificate of Analysis | 04/08/2022 | 041573.18,041573.30 |
Q07B035 | Certificate of Analysis | 07/12/2021 | 041573.18,041573.30 |
K16W045 | Certificate of Analysis | 07/12/2021 | 041573.18,041573.30 |
G03Z049 | Certificate of Analysis | 07/12/2021 | 041573.18,041573.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.