missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11373837

Antimony(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11373837
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11373837

Brand: Thermo Scientific Alfa Aesar 035676.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7790-44-5
Molecular Formula I3Sb
Molecular Weight (g/mol) 502.47
MDL Number MFCD00016322
InChI Key KWQLUUQBTAXYCB-UHFFFAOYSA-K
Synonym antimony iii iodide, stibine, triiodo, antimony triiodide, antimon iii iodid, antimony iodide sbi3, antimony iodide, antimony iii iodide, ultra dry, antimony iii triiodide, hsdb 437, triiodostibine #Show MoreShow Less
PubChem CID 24630
SMILES [Sb+3].[I-].[I-].[I-]

Especificaciones

Boiling Point 420°C
Melting Point 168°C
Physical Form Beads
Quantity 25 g
UN Number UN3260
Merck Index 14,710
Solubility Information Decomposed by water to SbOI.
IUPAC Name triiodostibane
Formula Weight 502.46
Percent Purity 99.998%
Odor Acrid
Packaging Ampule
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Antimony(III) iodide, Ultra dry
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT16E050 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number035676.03,035676.14
Lot NumberM20I037 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number035676.03,035676.14
Lot NumberE02L16 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number035676.03,035676.14
Lot NumberU31D029 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number035676.03,035676.14
Lot NumberT16E050 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number035676.03,035676.14

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Antimony(III) iodide, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado