missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) oxide, 99+%, Thermo Scientific Chemicals Product Code.: 11904161

Antimony(III) oxide, 99+%, Thermo Scientific Chemicals

Product Code. 11904161
5 kg
Click to view available options
Quantity:
100 g
1 kg
5 kg
This item is not returnable. View return policy

Product Code. 11904161

Brand: Thermo Scientific Acros 213470051

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
Synonym Antimony trioxide
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C
Melting Point 656°C
Quantity 5 kg
Assay Percent Range 99% min. (Sb2O3)
Linear Formula Sb2O3
Merck Index 15,699
Solubility Information Solubility in water: 2.7mg/l (20°C). Other solubilities: slightly soluble in hot water,soluble in conc. hcl,acetic acid,sulfuric acids,,solutions of alkali hydroxides
Formula Weight 291.42
Percent Purity ≥99%
Packaging Plastic Bottle
Chemical Name or Material Antimony(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0398158 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number213470010,213471000,213470051
Lot NumberA0402236 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number213470010,213471000,213470051
Lot NumberA0411899 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number213470010,213471000,213470051
Lot NumberA0406166 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number213470010,213471000,213470051
Lot NumberA0404913 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number213470010,213471000,213470051
compliance-icons
Product Identifier
  • Antimony(III) oxide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) oxide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.