missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11320839

Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11320839
5 g
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 11320839

Brand: Thermo Scientific Alfa Aesar 010641.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antimony(III) oxide finds use as flame retardant wherein it synergistically acts with halogenated materials. It is employed in opacifying glasses, ceramics and enamels. In the preparation of optical and ruby glass, antimony trioxide serves as a bubble remover. Further, it is a useful catalyst in the production of polyethylene terephthalate (PET plastic) and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-64-4
Molecular Formula O3Sb2
Molecular Weight (g/mol) 291.52
MDL Number MFCD00011214
InChI Key GHPGOEFPKIHBNM-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1550°C (sublimation)
Melting Point 656°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metals basis)
Solubility Information Very slightly soluble in water; soluble in potassium hydroxide,hydrochloricacid and acetic acid; insoluble in organic solvents.
Formula Weight 291.5
Odor Odorless
Density 5.67
Chemical Name or Material Antimony(III) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX15K037 Certificate TypeCertificate of Analysis Date13/11/2024 Catalog Number010641.06,010641.18,010641.22,010641.14
Lot NumberR02K016 Certificate TypeCertificate of Analysis Date14/05/2024 Catalog Number010641.06,010641.18,010641.22,010641.14
Lot NumberS07J059 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number010641.06,010641.18,010641.22,010641.14
Lot NumberY23I027 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number010641.06,010641.18,010641.22,010641.14
Lot NumberP25H039 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number010641.06,010641.18,010641.22,010641.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.