missing translation for 'onlineSavingsMsg'
Learn More
Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals Product Code.: 11371849

Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals

Product Code. 11371849
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11371849

Brand: Thermo Scientific Alfa Aesar 012521.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is manufactured for use in safety matches, military ammunition, explosives and fireworks. It also is used in the production of ruby-colored glass and in plastics as a flame retardant. percussion caps, camouflage paints, ruby glass

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
Synonym Antimony trisulfide
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Color Black
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.0% min (Metal basis)
Solubility Information Insoluble in water. Soluble in HCl with H2S evolution
Formula Weight 339.68
Odor Odorless
Appearance Black powder
Density 4.12-4.64g/cm3
Chemical Name or Material Antimony(III) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS28K028 Certificate TypeCertificate of Analysis Date25/07/2024 Catalog Number012521.A1,012521.30
Lot NumberZ11J025 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number012521.A1,012521.30
Lot NumberY10B020 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012521.A1,012521.30
Lot NumberT01B025 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012521.A1,012521.30
Lot NumberF19N04 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012521.A1,012521.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony(III) sulfide, 98% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.