Learn More
Antimony(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Beschrijving
Antimony Trisulfide is used in the production of pyrotechnics and as a yellow pigment for glass and porcelain. In Vacuum deposition. Used in preparation of Vermilion or yellow pigment, for antimony salts, percussion caps, camouflage paints, and in ruby glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specificatie
Specificatie
Melting Point | 550°C |
Physical Form | Pieces |
Quantity | 250 g |
UN Number | UN1549 |
Merck Index | 14,713 |
Solubility Information | Insoluble in water. Soluble in concentrated acids. |
Formula Weight | 339.68 |
Percent Purity | 99.9% |
Assay | (metals basis) |
Chemical Name or Material | Antimony(III) sulfide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P06J032 | Certificate of Analysis | 09/05/2023 | 040371.30 |
Y01I009 | Certificate of Analysis | 11/11/2022 | 040371.09,040371.18,040371.30 |
P02I044 | Certificate of Analysis | 12/04/2022 | 040371.09,040371.18,040371.30 |
T05C007 | Certificate of Analysis | 08/12/2021 | 040371.09,040371.18,040371.30 |
F03U017 | Certificate of Analysis | 07/12/2021 | 040371.09,040371.18,040371.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.