missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony trifluoride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 376.0°C |
Melting Point | 292.0°C |
Color | White to Gray |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Assay Percent Range | 97.5% min. (Iodimetry) |
Linear Formula | SbF3 |
Fieser | 05,526; 06,553 |
Merck Index | 15, 697 |
Solubility Information | Solubility in water: 3850g/L (20°C). Other solubilities: decomposes in water, insoluble in nh3 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0428720 | Certificate of Analysis | 02/10/2023 | 191245000,191240050,191241000 |
A0400090 | Certificate of Analysis | 25/08/2023 | 191245000,191240050,191241000 |
A0452104 | Certificate of Analysis | 02/05/2023 | 191245000,191241000 |
A0374519 | Certificate of Analysis | 13/12/2022 | 191245000,191240050,191241000 |
A0436020 | Certificate of Analysis | 11/11/2021 | 191245000,191240050,191241000 |
Safety and Handling



Product Identifier
- Antimony trifluoride
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Antimony trifluoride, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction