Learn More
Applied Biosystems™ SYBR™ Green Universal Master Mix
Everything needed for SYBR™ Green dye-based PCR amplification and detection in a convenient, single-tube format.
Brand: Applied Biosystems™ 4334973
2859.50 GBP valid until 2025-12-31
Use promo code "24271" to get your promotional price.
Includes: 2X mixture of SYBR Green I Dye, AmpliTaq Gold DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components.
Description
Applied Biosystems™ SYBR™ Green Universal Master Mix combines SYBR™ Green I dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1, and optimized buffer in the convenience of a single vial.
- SYBR™ Green I dye detects double-stranded DNA, so specific probes are not required
- AmpliTaq Gold™ DNA polymerase minimizes nonspecific product formation to achieve superior performance
- dUTP significantly reduces carryover contamination when used in conjunction with uracil-DNA glycosylase
- Proprietary buffer enhancements ensure performance and reliability
- Premixed components stored at 2° to 8°C significantly reduce assay setup time
Maximum Flexibility and Convenience:
- Applied Biosystems™ SYBR™ Green Universal Master Mix provides maximum flexibility at reduced cost because no target-specific TaqMan™ probes are required
- SYBR™ Green I dye is a double-stranded DNA binding dye that detects any double-stranded DNA generated during PCR
- The hot-start enzyme AmpliTaq Gold™ DNA Polymerase minimizes nonspecific product formation (including primer-dimers), yielding superior performance and sensitivity
- Passive Internal Reference 1 is provided to normalize non-PCR—related fluorescence fluctuations
- This minimizes well-to-well variability that can result from a variety of causes, such as pipetting error or sample evaporation
- SYBR™ Green I dye is ideal for target identification (screening assays) or when a limited number of assays is needed.
- Store at -20°C
Order Info
Shipping Condition: Wet Ice

Specifications
The 1 x 50 mL vials contain a 2X mixture of SYBR Green 1 Dye, AmpliTaq Gold™ DNA Polymerase, dNTPs with dUTP, Passive Reference 1 (ROX), and optimized buffer components. Sufficient reagents provided for 2000 reactions based on a 50 μl reaction volume. Store at -20°C. Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA). |
|
Tube | |
qPCR | |
Standard | |
SYBR Green | |
SYBR™ | |
1 x 50 mL | |
2000 Reactions of 50 μL | |
2X | |
DNA (Genomic), cDNA |
SYBR | |
High | |
AmpliTaq Gold DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System | |
ROX (Pre-mixed) | |
Real Time PCR SYBR Master Mix | |
Wet Ice | |
Gene Expression | |
2000 Reactions | |
1 x 50 mL |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2410165 | Certificate of Analysis | 30/10/2024 | 4334973,4334973_LITL18 PROMO |
2407164 | Certificate of Analysis | 06/08/2024 | 4334973,4334973_LITL18 PROMO |
2405163 | Certificate of Analysis | 04/06/2024 | 4334973,4334973_LITL18 PROMO |
2405162 | Certificate of Analysis | 15/05/2024 | 4334973,4334973_LITL18 PROMO |
2403159 | Certificate of Analysis | 14/04/2024 | 4334973,4334973_LITL18 PROMO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.