Learn More
Karl Fischer Aqualine™ Complete 5K, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Brand: Fisher Chemical K/2250R/15
100.17 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
View more versions of this product
Description
- Single-component volumetric reagent for high water content analysis of aldehydes and ketones
- Water equivalent: 5.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free

Specifications
Aqualine™ Complete 5K | |
Karl Fischer Reagent | |
111-90-0, 5683-33-0, 7446-09-5, 7553-56-2 | |
Amber glass bottle | |
81734 |
Colorless | |
1 L | |
Report mg of water/mL | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Aqualine Complete 5K
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.