Learn More
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series
Aura™ Particulate Respirator 9300+Gen3 Series is the next generation of 3M Aura Respirator: FFP protection combined with the original 3-panel, flat-fold design.
£13.82 - £2165.90
Specifications
Certifications/Compliance (Europe) EN 149:2001 + A1:2009, CE Approved
Color White
Design Type Flat-fold
Disposable Single-use
Material Polypropylene
View More Specs
Products 10
Product Code Brand Filter Type Assigned Protection Factor (APF) With Exhalation Valve Quantity Price Quantity & Availability  
Product Code Brand Filter Type Assigned Protection Factor (APF) With Exhalation Valve Quantity Price Quantity & Availability  
16260262
View Documents
3M™
7100138120
FFP2 10 Yes 5/Pk. N/A N/A N/A
17817603
View Documents Promotion Details
3M™
7100138120
FFP2 10 Yes 1/Pk.
£186.00
Pack of 40
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
16250262
View Documents
3M™
7100138121
FFP2 10 No 5/Pk. N/A N/A N/A
16270262
View Documents
3M™
7100138357
FFP3 20 Yes 5/Pk. N/A N/A N/A
16220262
View Documents Promotion Details
3M™
7100134314
FFP3 20 Yes 10/Pk.
£78.70
Pack of 10
In Stock
to see stock.
Add to basket
16200262
View Documents Promotion Details
3M™
7100134313
FFP2 10 Yes 10/Pk.
£34.35
Pack of 10
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
16290252
View Documents
3M™
7100134298
FFP2 10 No 20/Pk. N/A N/A N/A
16210262
View Documents
3M™
7100134311
FFP3 20 No 20/Pk. N/A N/A N/A
16230262
View Documents
3M™
7100139467
FFP2 10 Yes 330/Pk. N/A N/A N/A
16240262
View Documents
3M™
7100138716
FFP3 20 Yes 330/Pk. N/A N/A N/A
Description
Description
  • Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
  • 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort compared to 3M™ Aura™ 9300+ equivalent model
  • 3M™ Advanced Electret (filter) Media helps you breathe easier
  • Innovative tabs on the upper and lower panels simplify positioning and adjustment
  • Braided headbands are colour-coded (blue for FFP2, red for FFP3) for quick and easy identification
  • Embossed upper panel reduces eyewear fogging
  • Easy to store away with flat-fold design and individual wrapping
Specifications
Specifications
EN 149:2001 + A1:2009, CE Approved
Flat-fold
Polypropylene
Standard
Non-sterile
No
White
Single-use
General Purpose Particulate Respirator
Polyisoprene
Flat-fold
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series