missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices
GREENER_CHOICEBIOPHARMA

Ensure consistent performance and quality. Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices are constructed of clear polypropylene in 96-well racks. Available in nonsterile/sterile, non-filtered/filtered, wide-bore and bottom-labeled options with volumes ranging from 50 to 200μL.

£89.66 - £809.70

Specifications

Color Clear
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Packaging SBS Rack
View More Specs

Products 15
Product Code Brand Volume (Metric) Sterility Filtered Tip Style Price Quantity & Availability  
Product Code Brand Volume (Metric) Sterility Filtered Tip Style Price Quantity & Availability  
11346284
View Documents
Axygen™
EV-50-R
50 μL Non-sterile Non-filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10059499
BIOPHARMA
View Documents Promotion Details
Axygen™
EV-50-R
50 μL Non-sterile Non-filtered Automation, Round
£566.47
Pack of 4800
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
12736689
BIOPHARMA
View Documents Promotion Details
Axygen™
EVF-180-R-S
180 μL Sterile Filtered Automation, Round
£647.14
Pack of 4800
In Stock
to see stock.
Add to basket
 
11376284
View Documents
Axygen™
EVF-180-R-S
180 μL Sterile Filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11356284
View Documents
Axygen™
EV-50-R-S
50 μL Sterile Non-filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12746689
GREENER_CHOICEBIOPHARMA
View Documents
Axygen™
EVF-205-WB-R-S
205 μL Sterile Filtered Automation, Wide Bore
£809.70
Pack of 4800
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
12756689
BIOPHARMA
View Documents Promotion Details
Axygen™
EVF-50-R-S
50 μL Sterile Filtered Automation, Round
£647.14
Pack of 4800
In Stock
to see stock.
Add to basket
 
11386284
View Documents
Axygen™
EVF-50-R-S
50 μL Sterile Filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11366284
View Documents
Axygen™
EVF-100-R-S
100 μL Sterile Filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11336284
View Documents
Axygen™
EV-200-R-S
200 μL Sterile Non-filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12752836
BIOPHARMA
View Documents Promotion Details
Axygen™
EV-200-R-S
200 μL Sterile Non-filtered Automation, Round
£583.50
Pack of 4800
In Stock
to see stock.
Add to basket
 
10535950
BIOPHARMA
View Documents Promotion Details
Axygen™
EV-200-R
200 μL Non-sterile Non-filtered Automation, Round
£566.47
Pack of 4800
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
11326284
View Documents
Axygen™
EV-200-R
200 μL Non-sterile Non-filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10810501
BIOPHARMA
View Documents
Axygen™
EV-100-R
100 μL Non-sterile Non-filtered Automation, Round
£552.97
Pack of 4800
Estimated Shipment: 28-08-2025
to see stock.
Add to basket
 
11306284
View Documents
Axygen™
EV-100-R
100 μL Non-sterile Non-filtered Automation, Round This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available in a range of volumes: 50, 100, 180 and 200μL
  • Compatible with the following:
    • Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head
    • Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with HV96 head
    • Biotek™ Precision™ and Precision XS 8 or 12 head
    • Dynamic Devices Oasis LM 8/96-Channel-Standard Vol and 96-Channel-Low Vol head
Specifications

Specifications

Clear
Non-pyrogenic, used for stringent specifications under strict process controls
SBS Rack
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius, Zymark/Caliper RapidPlate, SciClone, Zephyr, BioTek Precision, BioTek Precision XS, Dynamic Devices Oasis LM
DNase, Rnase Free
Rack
50 Racks/Cs.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.