Learn More
Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Wide Bore
Designed to fit a wide variety of single and multi-channel pipettes
Brand: Axygen™ T-205-WB-C-R-S
321.61 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Wide-Bore
Order Info
Available in bulk, racked or stack rack formats.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
1 to 200 μL | |
Clear | |
DNase, RNase, and Pyrogen free | |
Genomic/Wide Orifice, Universal | |
Polypropylene |
Sterile | |
Autoclavable | |
Rack | |
Universal Fit Pipettor |