Learn More
Axygen™ Automation Tips for STARLine, Clear, CO-RE Style Liquid Level Sensing Tip, Sterile
GREENER_CHOICEBIOPHARMA
For complete compatibility and a consistent fit.
Brand:  Axygen™ HT-50-CBK-STK-S
 View more versions of this product
Product Code. 15763997
  • £843.49 / Pack of 11520
Estimated Shipment: 23-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Axygen brand automation tips are engineered to be compatible with original equipment manufacturers' tips, ensuring complete compatibility and a consistent fit. One of the widest product offerings combined with our engineering expertise stringent QA/QC enables you to get optimum performance no matter which automated liquid handler you are working with.
  • Compatible with Hamilton™ Microlab™ STAR, STARlet, STARplus and Nimbus™
  • Conductive black polypropylene
  • Certified DNase-/RNase-free, nonpyrogenic
  • With or without aerosol-resistant filters
  • Nonsterile or sterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
Sterile
DNase, RNase Free
Round
Polypropylene
Non-filtered
Black
Refill
Hamilton MICROLAB STAR, Hamilton MICROLAB STARlet, Hamilton MICROLAB STARplus
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Automation Tips for STARLine, Clear, CO-RE Style Liquid Level Sensing Tip, Sterile > 50μL tip working volume