Learn More
Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips
GREENER_CHOICEBIOPHARMA
768 tips for use with Tecan Evo MCA96 multi-channel arm
Brand:  Axygen™ EV-50-NTR-S
 View more versions of this product
Product Code. 15277668
  • £351.81 / Pack of 3840
Estimated Shipment: 11-09-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: MCA96
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
Orange
Nested tip rack
Tecan EVO MCA96
Polypropylene
Sterile
Round
40 Racks/Cs.
Tecan
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number21722022 Certificate TypeCertificate of Compliance Date12/04/2024 Catalog NumberEV-50-NTR-S
Lot Number20122027 Certificate TypeCertificate of Compliance Date12/04/2024 Catalog NumberEV-50-NTR-S
Special Offers
Special Offers
Product Content Correction
Product Title
Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips > Non-filtered, Nested, Sterile; 50 μL