Learn More
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels
Label curved frozen lab surfaces including tubes, vials and cryocanes with the ultra-thin B-492 polyester label material. Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170894
103.55 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 17811749
  • £109.00 / Pack of 240
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for laboratory use on curved surfaces including test tubes, vials and straw pipettes
  • Withstands chemicals, abrasions and extreme temperatures, including storage in liquid nitrogen
  • Made with a permanent acrylic adhesive for tight bonding onto smooth curved surfaces such as glass and polypropylene
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ FreezerBondz™ Ultra-thin Cryogenic Polyester Labels > Dimensions: 25.40 mm x 12.70 mm x 9.53 mm (Dia); Printer Compatibility: BMP51;BMP53;M511;M510; Color: Black on White