Learn More
Tocris Bioscience™ AZ 10397767
Potent CXCR2 antagonist
Brand:  Tocris Bioscience™ 5872/10
171.06 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18777686
  • £214.00 / 10mg
Explore more special offers
Description
Description
AZ 10397767 is a potent CXCR2 antagonist (IC50 = 1 nM); attenuates oxaliplatin -induced NF-κB activation, increases oxaliplatin cytotoxicity, and potentiates oxaliplatin-induced apoptosis in AIPC cells. Reduces the numbers of neutrophils infiltrating into tumors in both in vitro and in vivo models and delayed tumor growth. Orally bioavailable.
Specifications
Specifications
Antagonist
333742-63-5
400.88g/mol
Chemokine Receptor Antagonists
Chemokine Receptor
C15H14ClFN4O2S2
Store at -20°C