missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ 10397767
Potent CXCR2 antagonist
£214.00 - £816.00
Specifications
Product Type | Antagonist |
---|---|
Target Proteins | Chemokine Receptor |
CAS | 333742-63-5 |
Molecular Formula | C15H14ClFN4O2S2 |
Formula Weight | 400.88g/mol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18777686
|
Tocris Bioscience™
5872/10 |
10 mg |
£214.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18797936
|
Tocris Bioscience™
5872/50 |
50 mg |
£816.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent CXCR2 antagonist (IC50 = 1 nM); attenuates oxaliplatin -induced NF-κB activation, increases oxaliplatin cytotoxicity, and potentiates oxaliplatin-induced apoptosis in AIPC cells. Reduces the numbers of neutrophils infiltrating into tumors in both in vitro and in vivo models and delayed tumor growth. Orally bioavailable.Specifications
Antagonist | |
333742-63-5 | |
400.88g/mol | |
Chemokine Receptor Antagonists |
Chemokine Receptor | |
C15H14ClFN4O2S2 | |
Store at -20°C |
For Research Use Only