missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ 10397767
Potent CXCR2 antagonist
Brand: Tocris Bioscience™ 5872/50
652.25 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
AZ 10397767 is a potent CXCR2 antagonist (IC50 = 1 nM); attenuates oxaliplatin -induced NF-κB activation, increases oxaliplatin cytotoxicity, and potentiates oxaliplatin-induced apoptosis in AIPC cells. Reduces the numbers of neutrophils infiltrating into tumors in both in vitro and in vivo models and delayed tumor growth. Orally bioavailable.Specifications
Antagonist | |
333742-63-5 | |
400.88g/mol | |
Chemokine Receptor Antagonists |
Chemokine Receptor | |
C15H14ClFN4O2S2 | |
Store at -20°C | |
50 mg |
For Research Use Only