missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Gas Delivery Tube
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
Brand: Azlon™ DT002
This item is not returnable.
View return policy
Description
- Pre-bent rigid polypropylene delivery tubes
- Unbreakable in normal use
- Suitable for many types of gas collection experiments in schools and universities

Specifications
Polypropylene | |
11.41 in. | |
Many types of gas collection experiments in schools and universities | |
Bung Top Gas Delivery Tube |
0.23 in. | |
290 mm | |
135°C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Azlon™ Gas Delivery Tube > Double bent; bung top; Length: 290mm
Spot an opportunity for improvement?Share a Content Correction