Learn More
Azlon™ LDPE Printed Slope Shoulder Wash Bottles
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. Azlon™ LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.
Brand: Azlon™ WGF501
Description
Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. AzlonTM LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.
- Labeled wash bottles for easy identification of contents
- Slim design enables comfortable handling and enhanced ease of use
- Fitted with color-coded polypropylene closures
- Closures feature precision jet-tip aperture for fine control

Specifications
500 mL | |
For easy identification of contents | |
LDPE | |
White | |
Not Autoclavable | |
Round | |
Eau distillée | |
69 mm |
Translucent | |
Color-coded Polypropylene closure | |
5/PK | |
Yes | |
Narrow | |
187 mm | |
Polypropylene | |
Cap |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.