Learn More
Tocris Bioscience™ B I09
IRE1 endonuclease inhibitor; cell permeable
Brand:  Tocris Bioscience™ 6009/10
168.46 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18705909
  • £221.00 / 10mg
Explore more special offers
Description
Description
B I09 is an IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.
Specifications
Specifications
Beige
1607803-67-7
303.31g/mol
Store at -20°C
10 mg
Inhibitor
C16H17NO5
Soluble to 50mM in DMSO
Inhibits Rnase