missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ B I09

IRE1 endonuclease inhibitor; cell permeable

£221.00 - £850.00

Specifications

Color Beige
Product Type Inhibitor
CAS 1607803-67-7
Molecular Formula C16H17NO5
Formula Weight 303.31g/mol
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18705909
View Documents Promotion Details
Tocris Bioscience™
6009/10
10 mg
£221.00
10mg
Please to purchase this item. Need a web account? Register with us today!
18780498
View Documents
Tocris Bioscience™
6009/50
50 mg
£850.00
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.
Specifications

Specifications

Beige
1607803-67-7
303.31g/mol
Store at -20°C
Inhibitor
C16H17NO5
Soluble to 50mM in DMSO
Inhibits Rnase
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only